Learn More
Grainger Honeywell Primair 100 Bib Hood, Tychem QC, Universal Size, 1 ea
Catalog No. 502795040
Encompass_Preferred
The Primair 100 bibbed hood assembly provides respiratory protection in a variety of industrial environments. This loose-fitting hood features an integrated bib and is constructed from Tychem QC material, which is coated to offer protection against intermittent splash. Designed for compatibility with specific supplied air and powered air-purifying respiratory systems, the assembly includes a co-polyester visor and a manifold that directs air across the lens to help minimize fogging.
  • Provides an assigned protection factor (APF) of 1,000
  • Features universal sizing with triple adjustment and a ratchet suspension
  • Constructed from Tychem QC-coated spun bond polypropylene
  • Includes a 0.040 mil co-polyester visor
  • Latex-free construction
  • Compatible with specific PAPR and SAR systems
Catalog No. 50-279-5040 Supplier Grainger Supplier No. 1AAN8
The Primair 100 bibbed hood assembly provides respiratory protection in a variety of industrial environments. This loose-fitting hood features an integrated bib and is constructed from Tychem QC material, which is coated to offer protection against intermittent splash. Designed for compatibility with specific supplied air and powered air-purifying respiratory systems, the assembly includes a co-polyester visor and a manifold that directs air across the lens to help minimize fogging.
  • Provides an assigned protection factor (APF) of 1,000
  • Features universal sizing with triple adjustment and a ratchet suspension
  • Constructed from Tychem QC-coated spun bond polypropylene
  • Includes a 0.040 mil co-polyester visor
  • Latex-free construction
  • Compatible with specific PAPR and SAR systems