Learn More
Grainger NASCO AcidBasic Bib Trouser, PVC, Green, 1 ea
Catalog No. 502774672
Encompass_Preferred
These green PVC bib overalls are designed to provide enhanced safety and superior quality in environments requiring chemical splash resistance. Constructed for durability and heavy-duty use, the trousers offer a flexible design that maximizes worker range of motion while providing reliable resistance to water penetration.
  • Dielectrically welded seams ensure 100% waterproof protection.
  • Chemical splash resistant construction helps minimize burn injuries from potential exposure.
  • Non-conductive design utilizes all non-metallic hardware.
  • Limited flame resistance provides self-extinguishing capabilities within 2 seconds per ASTM D6413.
  • Heavy-duty material facilitates longevity and reduces the need for replacements.
Catalog No. 50-277-4672 Supplier Grainger Supplier No. 178T75
These green PVC bib overalls are designed to provide enhanced safety and superior quality in environments requiring chemical splash resistance. Constructed for durability and heavy-duty use, the trousers offer a flexible design that maximizes worker range of motion while providing reliable resistance to water penetration.
  • Dielectrically welded seams ensure 100% waterproof protection.
  • Chemical splash resistant construction helps minimize burn injuries from potential exposure.
  • Non-conductive design utilizes all non-metallic hardware.
  • Limited flame resistance provides self-extinguishing capabilities within 2 seconds per ASTM D6413.
  • Heavy-duty material facilitates longevity and reduces the need for replacements.