Learn More
Enzo Life Sciences BIBB-515 (50mg). CAS: 156635-05-1
SDP
Supplier:  Enzo Life Sciences BMLEI3400050
Encompass_Preferred
BIBB 515 is a potent and selective inhibitor of 2,3-oxidosqualene cyclase (OSC) in vivo. BIBB 515 inhibits LDL production in both normolipemic and hyperlipemic hamsters and may represent a novel approach for the development of lipid lowering drugs. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (2mg/ml) or DMF (8mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0724