Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bibenzyl, 99%

Catalog No. p-67044
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106230050 5 g Glass bottle
AC106230250 25 g Glass bottle
AC106231000 100 g Glass bottle
AC106235000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC106230050 Supplier Thermo Scientific Chemicals Supplier No. 106230050
Only null left
Add to Cart

CAS: 103-29-7 | C14H14 | 182.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-29-7
Molecular Formula C14H14
Molecular Weight (g/mol) 182.27
MDL Number MFCD00004796
InChI Key QWUWMCYKGHVNAV-UHFFFAOYSA-N
Synonym 1,2-diphenylethane, bibenzyl, dibenzyl, s-diphenylethane, 2-phenylethyl benzene, 1,2-dihydrostilbene, dibenzil, sym-diphenylethane, dihydrostilbene, benzene, 1,1'-1,2-ethanediyl bis
PubChem CID 7647
ChEBI CHEBI:34047
IUPAC Name 2-phenylethylbenzene
SMILES C(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 49.0°C to 53.0°C
Color White to Yellow
Density 1.0100g/mL
Boiling Point 284.0°C
Flash Point 129°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH2C6H5
Quantity 5 g
Beilstein 05,598
Specific Gravity 1.01
Merck Index 15,1202
Solubility Information Solubility in water: practically insoluble. Other solubilities: practically insoluble in liquid ammonia, moderately soluble in alcohol, freely soluble in carbon disulfide, ether,, chloroform and amyl acetate, soluble in liquid sulfur dioxide
Formula Weight 182.26
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Bibenzyl, 99%
Show More Show Less
EINECSNumber 203-096-4
RTECSNumber DT4375000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.