Learn More
Cayman Chemical BIBF 1120-13C-d3, Purity: ≥99% deuterated forms (d1-d3), 1 mg
SDP
Catalog No. 502932501
Encompass_Preferred
An internal standard for the quantification of BIBF 1120 by GC- or LC-MS
Catalog No. 50-293-2501 Supplier Cayman Chemical Supplier No. 253121 MG
An internal standard for the quantification of BIBF 1120 by GC- or LC-MS