Learn More
Cayman Chemical BIBF 1120-13C-d3 500ug
SDP
Catalog No. 502932502
Encompass_Preferred
An internal standard for the quantification of BIBF 1120 by GC- or LC-MS
Catalog No. 50-293-2502 Supplier Cayman Chemical Supplier No. 25312500 UG
An internal standard for the quantification of BIBF 1120 by GC- or LC-MS