Learn More
Cayman Chemical BIBF 1120 (esylate), Purity: ≥98%, CAS: 656247-18-6, 100 mg
SDP
Catalog No. 502989729
Encompass_Preferred
A VEGFR, FGFR, and PDGFR inhibitor (IC50s = 13-34, 37-610, 59, and 65 nM for VEGFR1-3, FGFR1-4, PDGFRα, and PDGFRβ, respectively); selective for VEGFR, FGFR, and PDGFR over a panel of 33 kinases but does inhibit FLT3, LCK, LYN, and Src (IC50s = 16-156 nM); inhibits growth factor-dependent proliferation of HUVECs, HSMECs, HUASMCs, and BRPs (EC50s = 7-290 nM); reduces tumor microvessel density and the number of PDGFRβ-expressing perivascular cells in a FaDu head and neck small cell carcinoma mouse xenograft model at 100 mg/kg; inhibits tumor growth in a Caki-1 renal cancer mouse xenograft model
Catalog No. 50-298-9729 Supplier Cayman Chemical Supplier No. 31082100 MG
A VEGFR, FGFR, and PDGFR inhibitor (IC50s = 13-34, 37-610, 59, and 65 nM for VEGFR1-3, FGFR1-4, PDGFRα, and PDGFRβ, respectively); selective for VEGFR, FGFR, and PDGFR over a panel of 33 kinases but does inhibit FLT3, LCK, LYN, and Src (IC50s = 16-156 nM); inhibits growth factor-dependent proliferation of HUVECs, HSMECs, HUASMCs, and BRPs (EC50s = 7-290 nM); reduces tumor microvessel density and the number of PDGFRβ-expressing perivascular cells in a FaDu head and neck small cell carcinoma mouse xenograft model at 100 mg/kg; inhibits tumor growth in a Caki-1 renal cancer mouse xenograft model