Learn More
Apexbio Technology LLC BIBF 1202 894783-71-2 5mg
SDP
Catalog No. 5000097160
Encompass_Preferred
BIBF 1202 (CAS 894783-71-2) is a carboxylate metabolite derivative of BIBF 1120 characterized as a selective inhibitor of vascular endothelial growth factor receptor 2 (VEGFR2) kinase In biochemical assays BIBF 1202 demonstrates inhibitory activity against VEGFR2 kinase with an IC50 of 62 nM This compound is frequently employed in studies investigating the regulation of the VEGFR2 signaling pathway and its role in endothelial cell function Additionally BIBF 1202 is utilized in preclinical models related to tumor angiogenesis and associated pharmacological research
Catalog No. 50-000-97160 Supplier Apexbio Technology LLC Supplier No. A46945
BIBF 1202 (CAS 894783-71-2) is a carboxylate metabolite derivative of BIBF 1120 characterized as a selective inhibitor of vascular endothelial growth factor receptor 2 (VEGFR2) kinase In biochemical assays BIBF 1202 demonstrates inhibitory activity against VEGFR2 kinase with an IC50 of 62 nM This compound is frequently employed in studies investigating the regulation of the VEGFR2 signaling pathway and its role in endothelial cell function Additionally BIBF 1202 is utilized in preclinical models related to tumor angiogenesis and associated pharmacological research