Learn More
TARGETMOL CHEMICALS INC BIBF0775 5MG
SDP
Catalog No. 502579795
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BIBF0775 is a selective TGF(beta) type I receptor (Alk5) inhibitor (IC50 34 nM). purity: 99%
Catalog No. 50-257-9795 Supplier TARGETMOL CHEMICALS INC Supplier No. T51975MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BIBF0775 is a selective TGF(beta) type I receptor (Alk5) inhibitor (IC50 34 nM). purity: 99%