Learn More
Selleck Chemical LLC Nintedanib (BIBF 1120) 5mg 656247-17-5 Intedanib, Vargatef, Ofev
SDP
Catalog No. 506877
Encompass_Preferred
Nintedanib (BIBF 1120, Intedanib, Vargatef, Ofev) is a potent triple angiokinase inhibitor for VEGFR1/2/3, FGFR1/2/3 and PDGFR?/? with IC50 of 34 nM/13 nM/13 nM, 69 nM/37 nM/108 nM and 59 nM/65 nM in cell-free assays. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-687-7 Supplier Selleck Chemical LLC Supplier No. S10105MG
Nintedanib (BIBF 1120, Intedanib, Vargatef, Ofev) is a potent triple angiokinase inhibitor for VEGFR1/2/3, FGFR1/2/3 and PDGFR?/? with IC50 of 34 nM/13 nM/13 nM, 69 nM/37 nM/108 nM and 59 nM/65 nM in cell-free assays. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.