Learn More
Medchemexpress LLC Bibo 3304 trifluoroacetate | 191868-14-1 | MFCD04112989 | 100.0% | 643.66 g/mol | C31H36F3N7O5 | 5 MG
SDP
Catalog No. 5000241201
Encompass_Preferred
BIBO3304 (trifluoroacetate) is a potent, orally active, and selective neuropeptide Y (NPY) Y1 receptor antagonist used as a research tool to probe NPY1R-mediated signaling in pharmacology and neuroscience studies. It is supplied as a high-purity solid and is characterized for receptor binding and in vitro functional assays.
  • High potency and selectivity for the NPY Y1 receptor.
  • Suitable for in vitro receptor binding and functional assays.
  • High reported purity for reproducible experimental results.
  • Available as a solid and as a DMSO solution for assay convenience.
  • Well characterized with supporting chemical identifiers and structural data.
Catalog No. 50-002-41201 Supplier Medchemexpress LLC Supplier No. HY1077255MG
BIBO3304 (trifluoroacetate) is a potent, orally active, and selective neuropeptide Y (NPY) Y1 receptor antagonist used as a research tool to probe NPY1R-mediated signaling in pharmacology and neuroscience studies. It is supplied as a high-purity solid and is characterized for receptor binding and in vitro functional assays.
  • High potency and selectivity for the NPY Y1 receptor.
  • Suitable for in vitro receptor binding and functional assays.
  • High reported purity for reproducible experimental results.
  • Available as a solid and as a DMSO solution for assay convenience.
  • Well characterized with supporting chemical identifiers and structural data.