Learn More
Selleck Chemical LLC BIBO 3304 Trifluoroacetate
SDP
Catalog No. 503134774
Encompass_Preferred
BIBO 3304 Trifluoroacetate (BIBO-3304 TFA) is a novel neuropeptide Y Y1 receptor (NPY1R) antagonist with IC50 values of 0 38 nM and 0 72 nM for human Y1 receptor and rat Y1 receptor respectively BIBO 3304 shows low affinity for the human Y2 receptor human and rat Y4 receptor as well as for the human and rat Y5 receptor (IC50 values 1000 nM)
Catalog No. 50-313-4774 Supplier Selleck Chemical LLC Supplier No. E001425MG
BIBO 3304 Trifluoroacetate (BIBO-3304 TFA) is a novel neuropeptide Y Y1 receptor (NPY1R) antagonist with IC50 values of 0 38 nM and 0 72 nM for human Y1 receptor and rat Y1 receptor respectively BIBO 3304 shows low affinity for the human Y2 receptor human and rat Y4 receptor as well as for the human and rat Y5 receptor (IC50 values 1000 nM)