Learn More
Cayman Chemical BIBO-3304 (trifluoroacetate salt), Purity: ≥98%, CAS: 191868-14-1, 10 mg
SDP
Catalog No. 502910391
Encompass_Preferred
An antagonist of NPY receptor Y1 (Kis = 0.38 and 0.72 nM, respectively); selective for Y1 over Y2, Y4, and Y5 receptors (Kis = >1,000 nM for all); inhibits NPY-induced increases in food intake in fasted rats when injected into the PVN of the hypothalamus at 30 µg/animal; inhibits NPY- or bilateral common carotid artery occlusion-induced pressor response in cats at 100 µg/kg
Catalog No. 50-291-0391 Supplier Cayman Chemical Supplier No. 3520510 MG
An antagonist of NPY receptor Y1 (Kis = 0.38 and 0.72 nM, respectively); selective for Y1 over Y2, Y4, and Y5 receptors (Kis = >1,000 nM for all); inhibits NPY-induced increases in food intake in fasted rats when injected into the PVN of the hypothalamus at 30 µg/animal; inhibits NPY- or bilateral common carotid artery occlusion-induced pressor response in cats at 100 µg/kg