Learn More
Medchemexpress LLC Bibo3304 | 191868-14-1 | 100.0% | 643.66 | C31H36F3N7O5 | 25 MG
SDP
Catalog No. 5000242758
Encompass_Preferred
BIBO3304 is a potent, selective neuropeptide Y (NPY) Y1 receptor antagonist used as a research probe. It exhibits subnanomolar affinity at human and rat Y1 receptors (IC50 = 0.38 nM and 0.72 nM) and is supplied in solid and solution formats with high reported purity.
  • High potency with subnanomolar IC50 values for human and rat Y1 receptors.
  • High reported purity (99.96%).
  • Available as solid and as a 10 mM DMSO solution for assay readiness.
  • Molecular formula C31H36F3N7O5 and molecular weight 643.66 g/mol.
  • Suitable for receptor pharmacology and in vitro pharmacodynamic studies.
Catalog No. 50-002-42758 Supplier Medchemexpress LLC Supplier No. HY10772525MG
BIBO3304 is a potent, selective neuropeptide Y (NPY) Y1 receptor antagonist used as a research probe. It exhibits subnanomolar affinity at human and rat Y1 receptors (IC50 = 0.38 nM and 0.72 nM) and is supplied in solid and solution formats with high reported purity.
  • High potency with subnanomolar IC50 values for human and rat Y1 receptors.
  • High reported purity (99.96%).
  • Available as solid and as a 10 mM DMSO solution for assay readiness.
  • Molecular formula C31H36F3N7O5 and molecular weight 643.66 g/mol.
  • Suitable for receptor pharmacology and in vitro pharmacodynamic studies.