Learn More
Medchemexpress LLC Bibo3304 trifluoroacetate | 191868-14-1 | MFCD04112989 | 100.0% | 643.66 g·mol⁻¹ | C31H36F3N7O5 | 10 MG
SDP
Catalog No. 5000242352
Encompass_Preferred
BIBO3304 is a potent, selective neuropeptide Y (NPY) Y1 receptor antagonist used in pharmacological and receptor biology research. Supplied as a trifluoroacetate salt in solid form, it is characterized by a defined molecular formula and molecular weight and is intended for in vitro and in vivo experimental applications.
  • Potent NPY Y1 receptor antagonist with subnanomolar affinity.
  • High reported purity suitable for research-grade assays.
  • Stable solid trifluoroacetate salt for convenient handling and storage.
  • Available in small milligram quantities and pre-made DMSO solutions for assay readiness.
  • Characterized molecular weight and formula to support analytical and dosing calculations.
Catalog No. 50-002-42352 Supplier Medchemexpress LLC Supplier No. HY10772510MG
BIBO3304 is a potent, selective neuropeptide Y (NPY) Y1 receptor antagonist used in pharmacological and receptor biology research. Supplied as a trifluoroacetate salt in solid form, it is characterized by a defined molecular formula and molecular weight and is intended for in vitro and in vivo experimental applications.
  • Potent NPY Y1 receptor antagonist with subnanomolar affinity.
  • High reported purity suitable for research-grade assays.
  • Stable solid trifluoroacetate salt for convenient handling and storage.
  • Available in small milligram quantities and pre-made DMSO solutions for assay readiness.
  • Characterized molecular weight and formula to support analytical and dosing calculations.