Learn More
Medchemexpress LLC BIBP3226 TFA | 1068148-47-9 | 99.3% | 587.59 | 5 MG
SDP
Catalog No. 5000364446
Encompass_Preferred
BIBP3226 TFA is a potent and selective neuropeptide Y Y1 (NPY Y1) and neuropeptide FF (NPFF) receptor antagonist, with Ki values of 1.1, 79, and 108 nM for rNPY Y1, hNPFF2, and rNPFF, respectively. It has been observed to display an anxiogenic-like effect.
  • Potent and selective neuropeptide Y Y1 (NPY Y1) and neuropeptide FF (NPFF) receptor antagonist.
  • Ki values of 1.1 nM for rNPY Y1, 79 nM for hNPFF2, and 108 nM for rNPFF.
  • Induces an anxiogenic-like effect at higher doses in vivo.
  • Available in 5 mg quantity.
Catalog No. 50-003-64446 Supplier Medchemexpress LLC Supplier No. 5MGHY107726
BIBP3226 TFA is a potent and selective neuropeptide Y Y1 (NPY Y1) and neuropeptide FF (NPFF) receptor antagonist, with Ki values of 1.1, 79, and 108 nM for rNPY Y1, hNPFF2, and rNPFF, respectively. It has been observed to display an anxiogenic-like effect.
  • Potent and selective neuropeptide Y Y1 (NPY Y1) and neuropeptide FF (NPFF) receptor antagonist.
  • Ki values of 1.1 nM for rNPY Y1, 79 nM for hNPFF2, and 108 nM for rNPFF.
  • Induces an anxiogenic-like effect at higher doses in vivo.
  • Available in 5 mg quantity.