Learn More
Apexbio Technology LLC BIBP 3226 trifluoroacetate(Synonyms: BIBP3226 TFA, BIBP3226 trifluoroacetate salt, NPY Y1 receptor antagonist BIBP3226), 2mg, CAS: 1068148-47-9.
SDP
Catalog No. 5000060835
Encompass_Preferred
BIBP 3226 trifluoroacetate (CAS 1068148-47-9) is a non-peptide antagonist targeting neuropeptide Y Y1 (NPY Y1) and neuropeptide FF (NPFF) receptors showing binding affinity with Ki values of 1 1 nM (rNPY Y1) 79 nM (hNPFF2) and 108 nM (rNPFF) BIBP 3226 competes with NPFF prevents NPFF-induced inhibition of forskolin-stimulated cAMP production and blocks NPFF-dependent hypothermic and anti-opioid effects in rodent models This compound is employed experimentally to investigate NPY/NPFF system roles in anxiety analgesia and cardiovascular regulation
Catalog No. 50-000-60835 Supplier Apexbio Technology LLC Supplier No. B71552
BIBP 3226 trifluoroacetate (CAS 1068148-47-9) is a non-peptide antagonist targeting neuropeptide Y Y1 (NPY Y1) and neuropeptide FF (NPFF) receptors showing binding affinity with Ki values of 1 1 nM (rNPY Y1) 79 nM (hNPFF2) and 108 nM (rNPFF) BIBP 3226 competes with NPFF prevents NPFF-induced inhibition of forskolin-stimulated cAMP production and blocks NPFF-dependent hypothermic and anti-opioid effects in rodent models This compound is employed experimentally to investigate NPY/NPFF system roles in anxiety analgesia and cardiovascular regulation