Learn More
Medchemexpress LLC BIBP3226 trifluoroacetate | 1068148-47-9 | MFCD168148479 | 99.3% | 587.59 | C29H32F3N5O5 | 10 MG
SDP
Catalog No. 5000241203
Encompass_Preferred
This compound is the trifluoroacetate (TFA) salt of a potent, selective non-peptide antagonist of neuropeptide Y Y1 and neuropeptide FF receptors. It is supplied as a dry research reagent suitable for receptor pharmacology and in vivo behavioral studies, reported to be soluble in DMSO and provided with high analytical purity.
  • Potent, selective antagonist of NPY Y1 and NPFF receptors.
  • Trifluoroacetate salt form for improved stability and handling.
  • High purity (reported 99.3%).
  • Soluble in DMSO for easy formulation in assays.
  • Available in multiple small package sizes for research use (5-500 mg).
Catalog No. 50-002-41203 Supplier Medchemexpress LLC Supplier No. HY10772610MG
This compound is the trifluoroacetate (TFA) salt of a potent, selective non-peptide antagonist of neuropeptide Y Y1 and neuropeptide FF receptors. It is supplied as a dry research reagent suitable for receptor pharmacology and in vivo behavioral studies, reported to be soluble in DMSO and provided with high analytical purity.
  • Potent, selective antagonist of NPY Y1 and NPFF receptors.
  • Trifluoroacetate salt form for improved stability and handling.
  • High purity (reported 99.3%).
  • Soluble in DMSO for easy formulation in assays.
  • Available in multiple small package sizes for research use (5-500 mg).