Learn More
Medchemexpress LLC HY-W004360 10mg Medchemexpress, BIBR 1087 SE CAS:212321-78-3 Purity:>98%
SDP
Catalog No. 501934482
Encompass_Preferred
Medchemexpress, HY-W004360 10mg BIBR 1087 SE CAS:212321-78-3 BIBR 1087 SE is an intermediate metabolite of dabigatran etexilate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4482 Supplier Medchemexpress LLC Supplier No. HYW00436010MG
Medchemexpress, HY-W004360 10mg BIBR 1087 SE CAS:212321-78-3 BIBR 1087 SE is an intermediate metabolite of dabigatran etexilate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.