Learn More
Medchemexpress LLC BIBR 1087 SE (Desethyl Dabigatran Etexilate) | 212321-78-3 | 599.68 | 10 MM * 1 ML
SDP
Catalog No. 5000446956
Encompass_Preferred
BIBR 1087 SE is an intermediate metabolite of dabigatran etexilate. This compound is provided as a white to yellow solid.
  • For research use only
  • In vitro solubility in DMSO is 250 mg/mL
  • Stable as powder at -20°C for 3 years and 4°C for 2 years
  • Stable in solvent at -80°C for 6 months and -20°C for 1 month
Catalog No. 50-004-46956 Supplier Medchemexpress LLC Supplier No. HYW00436010MM/1ML
BIBR 1087 SE is an intermediate metabolite of dabigatran etexilate. This compound is provided as a white to yellow solid.
  • For research use only
  • In vitro solubility in DMSO is 250 mg/mL
  • Stable as powder at -20°C for 3 years and 4°C for 2 years
  • Stable in solvent at -80°C for 6 months and -20°C for 1 month