Learn More
Selleck Chemical LLC BIBR 1532 10mg 321674-73-1
SDP
Supplier:  Selleck Chemical LLC S118610MG
Encompass_Preferred
BIBR 1532 is a potent, selective, non-competitive telomerase inhibitor with IC50 of 100 nM in a cell-free assay. No inhibition of DNA and RNA polymerases, including HIV reverse transcriptase are observed at concentrations vastly exceeding the IC50 for telomerase. BIBR 1532 induces apoptosis in cancer cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-887-6