Learn More
Selleck Chemical LLC Dabigatran (BIBR 953) 50mg 211914-51-1
SDP
Supplier:  Selleck Chemical LLC S219650MG
Encompass_Preferred
Dabigatran (BIBR 953) is a potent nonpeptide thrombin inhibitor with an IC50 of 9.3 nM in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-534-7