Learn More
Medchemexpress LLC BIBS 39 | 133085-33-3 | 99.7% | 524.65 | 1 MG
SDP
Catalog No. 5000400976
Encompass_Preferred
BIBS 39 is a novel nonpeptide angiotensin II (AII) receptor antagonist, primarily used for research. It demonstrates high selectivity for the AT1 subtype receptor compared to the AT2 subtype, with a Ki value of 29 ± 7 nM for AT1 and 480 ± 110 nM for AT2. This compound acts as a potent antagonist of the cardiovascular effects of AII.
  • Nonpeptide angiotensin II (AII) receptor antagonist
  • High selectivity for AT1 subtype
  • Potent antagonist of cardiovascular effects of AII
  • Suitable for research applications
  • Exhibits affinity for both AT1 and AT2 receptors
Catalog No. 50-004-00976 Supplier Medchemexpress LLC Supplier No. HY197321MG
BIBS 39 is a novel nonpeptide angiotensin II (AII) receptor antagonist, primarily used for research. It demonstrates high selectivity for the AT1 subtype receptor compared to the AT2 subtype, with a Ki value of 29 ± 7 nM for AT1 and 480 ± 110 nM for AT2. This compound acts as a potent antagonist of the cardiovascular effects of AII.
  • Nonpeptide angiotensin II (AII) receptor antagonist
  • High selectivity for AT1 subtype
  • Potent antagonist of cardiovascular effects of AII
  • Suitable for research applications
  • Exhibits affinity for both AT1 and AT2 receptors