Learn More
Cayman Chemical Bicalutamide, Purity: ≥98%, CAS: 90357-06-5, 100 mg
SDP
Catalog No. 502955869
Encompass_Preferred
A non-steroidal antiandrogen that binds the androgen receptor (Ki = 12.5 μM; IC50 = 1.2 μM), preventing its activation and subsequent upregulation of androgen responsive genes by androgenic hormones
Catalog No. 50-295-5869 Supplier Cayman Chemical Supplier No. 14250100 MG
A non-steroidal antiandrogen that binds the androgen receptor (Ki = 12.5 μM; IC50 = 1.2 μM), preventing its activation and subsequent upregulation of androgen responsive genes by androgenic hormones