Learn More
Enzo Life Sciences Bicalutamide (100mg). CAS: 90357-06-5
SDP
Supplier:  Enzo Life Sciences ALX270476M100
Encompass_Preferred
Nonsteroidal antiandrogen. Effective in the treatment of prostate cancer. Induces cell death by a pathway that is independent of changes in mitochondrial membrane potential and Bcl-2 action. Alternative name: N-(4-cyano-3-(trifluoromethyl)phenyl)-3-((4-fluorophenyl)sulfonyl)-2-hydroxy-2-methylpropanamide. Purity: ≥98% (HPLC). Appearance: White crystalline powder. Solubility: Soluble in acetone and tetrahydrofuran; slightly soluble in 100% ethanol or methanol; practically insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8602