Learn More
Cayman Chemical Bicalutamide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1185035-71-5, 1 mg
SDP
Catalog No. 502987800
Encompass_Preferred
An internal standard for the quantification of bicalutamide by GC- or LC-MS
Catalog No. 50-298-7800 Supplier Cayman Chemical Supplier No. 223771 MG
An internal standard for the quantification of bicalutamide by GC- or LC-MS