Learn More
Cayman Chemical BIcalutamIde-d4 500ug
SDP
Catalog No. 502987762
Encompass_Preferred
An internal standard for the quantification of bicalutamide by GC- or LC-MS
Catalog No. 50-298-7762 Supplier Cayman Chemical Supplier No. 22377500 UG
An internal standard for the quantification of bicalutamide by GC- or LC-MS