Learn More
Selleck Chemical LLC Bicalutamide 50mg 90357-06-5 ICI-176334
SDP
Catalog No. 506239
Encompass_Preferred
Bicalutamide (ICI-176334) is an androgen receptor (AR) antagonist with IC50 of 0.16 ?M in LNCaP/AR(cs)cell line. Bicalutamide promotes autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-623-9 Supplier Selleck Chemical LLC Supplier No. S119050MG
Bicalutamide (ICI-176334) is an androgen receptor (AR) antagonist with IC50 of 0.16 ?M in LNCaP/AR(cs)cell line. Bicalutamide promotes autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.