Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicalutamide, Tocris Bioscience™

Catalog No. 338950
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-895-0 50 mg
33-891-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-895-0 Supplier R&D Systems Supplier No. 3389/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Non-steroidal androgen receptor antagonist

Orally active non-steroidal androgen receptor antagonist (IC50 = 190 nM). Displays peripheral selectivity and does not effect serum levels of LH and testosterone. Exhibits potent anticancer activity in vivo.

Chemical Identifiers

CAS 90357-06-5
Molecular Formula C18H14F4N2O4S
Molecular Weight (g/mol) 430.374
InChI Key LKJPYSCBVHEWIU-UHFFFAOYSA-N
Synonym bicalutamide, casodex, cosudex, calutide, bicalutamide cdx, raffolutil, kalumid, casodex tn, bicalutamide usan:inn:ban, bicalutamide casodex
PubChem CID 2375
IUPAC Name N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide
SMILES CC(CS(=O)(=O)C1=CC=C(C=C1)F)(C(=O)NC2=CC(=C(C=C2)C#N)C(F)(F)F)O

Specifications

Quantity 50 mg
Formula Weight 430.37
Percent Purity >99%
Chemical Name or Material Bicalutamide
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.