Learn More
Medchemexpress LLC Bicarbonate-13C sodium | 87081-58-1 | MFCD00084061 | 98.0% | 85.00 | 13CHNaO3 | 50 MG
SDP
Catalog No. 5000281210
Encompass_Preferred
Bicarbonate-13C sodium is the 13C-enriched form of sodium bicarbonate supplied for laboratory and analytical use. It functions as an inorganic buffering agent and a stable isotope tracer for metabolic, biochemical, and NMR applications. Handle and store under dry conditions to minimize decomposition and preserve isotopic integrity.
  • 13C-enriched bicarbonate for isotope tracing and NMR studies
  • Provides pH buffering and culture medium stabilization
  • Applicable to biochemical assays and metabolic experiments
  • Hygroscopic; store sealed and protect from moisture
  • Provided in small research pack sizes suitable for laboratory use
Catalog No. 50-002-81210 Supplier Medchemexpress LLC Supplier No. HYW251598S50MG
Bicarbonate-13C sodium is the 13C-enriched form of sodium bicarbonate supplied for laboratory and analytical use. It functions as an inorganic buffering agent and a stable isotope tracer for metabolic, biochemical, and NMR applications. Handle and store under dry conditions to minimize decomposition and preserve isotopic integrity.
  • 13C-enriched bicarbonate for isotope tracing and NMR studies
  • Provides pH buffering and culture medium stabilization
  • Applicable to biochemical assays and metabolic experiments
  • Hygroscopic; store sealed and protect from moisture
  • Provided in small research pack sizes suitable for laboratory use