Learn More
Medchemexpress LLC Bicarbonate-13C sodium | 87081-58-1 | MFCD00084061 | 98.0% | 85.00 g/mol | NaH(13C)O3 | 500 MG
SDP
Catalog No. 5000280606
Encompass_Preferred
13C-labeled sodium bicarbonate is the isotopically labeled form of sodium bicarbonate used as a tracer and pH buffer in biochemical and metabolic studies. Provided as a solid with 98.0% purity (CAS 87081-58-1; formula NaH(13C)O3; MW 85.00 g/mol), it is suitable for buffering culture media, isotope tracing, and analytical applications. Product documentation includes a datasheet and safety data sheet for handling.
  • High isotopic enrichment for tracer experiments.
  • 98.0% purity for reproducible results.
  • Solid form for easy storage and handling.
  • Suitable for pH buffering in biological media.
  • Includes datasheet and safety data sheet for laboratory use.
Catalog No. 50-002-80606 Supplier Medchemexpress LLC Supplier No. HYW251598S500MG
13C-labeled sodium bicarbonate is the isotopically labeled form of sodium bicarbonate used as a tracer and pH buffer in biochemical and metabolic studies. Provided as a solid with 98.0% purity (CAS 87081-58-1; formula NaH(13C)O3; MW 85.00 g/mol), it is suitable for buffering culture media, isotope tracing, and analytical applications. Product documentation includes a datasheet and safety data sheet for handling.
  • High isotopic enrichment for tracer experiments.
  • 98.0% purity for reproducible results.
  • Solid form for easy storage and handling.
  • Suitable for pH buffering in biological media.
  • Includes datasheet and safety data sheet for laboratory use.