Learn More
Medchemexpress LLC Bicarbonate-13C sodium | 87081-58-1 | MFCD00084061 | 98.0% | 85.00 g/mol | NaH13CO3 | 250 MG
SDP
Catalog No. 5000281204
Encompass_Preferred
Bicarbonate-13C sodium is a 13C-labeled sodium bicarbonate supplied as a dry inorganic salt for use as a tracer and internal standard in analytical and metabolic studies. It is applicable to NMR, GC-MS, and LC-MS quantitation, and is useful for pH buffering in biological experiments.
  • 13C-labeled tracer for metabolic and pH imaging studies.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • High chemical purity (≥98.0%).
  • Stable solid form for straightforward storage and handling.
  • Useful for buffering and pH control in biological media.
Catalog No. 50-002-81204 Supplier Medchemexpress LLC Supplier No. HYW251598S250MG
Bicarbonate-13C sodium is a 13C-labeled sodium bicarbonate supplied as a dry inorganic salt for use as a tracer and internal standard in analytical and metabolic studies. It is applicable to NMR, GC-MS, and LC-MS quantitation, and is useful for pH buffering in biological experiments.
  • 13C-labeled tracer for metabolic and pH imaging studies.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • High chemical purity (≥98.0%).
  • Stable solid form for straightforward storage and handling.
  • Useful for buffering and pH control in biological media.