Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bicarbonate-13C sodium | 87081-58-1 | MFCD00084061 | 98.0% | 85.00 | NaH13CO3 | 100 MG
SDP

Catalog No. 5000280875
Encompass_Preferred

Bicarbonate-13C sodium is the 13C-isotopically labeled form of sodium bicarbonate used as a tracer, labeled standard, and reagent in metabolic and analytical studies. It is supplied as a dry powder with high chemical purity and defined isotopic enrichment for quantitative experiments.

  • Purity ≥98.0%.
  • Isotopically labeled at the bicarbonate carbon (13C).
  • Molecular formula NaH13CO3, molecular weight 85.00.
  • Available in small-scale packages including 100 mg for analytical use.
  • Recommended storage: sealed, away from moisture and light at 4°C; in solvent: -80°C up to 6 months.
  • Suitable for tracer studies, metabolic labeling, and analytical standards.

Catalog No. 50-002-80875 Supplier Medchemexpress LLC Supplier No. HYW251598S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bicarbonate-13C sodium is the 13C-isotopically labeled form of sodium bicarbonate used as a tracer, labeled standard, and reagent in metabolic and analytical studies. It is supplied as a dry powder with high chemical purity and defined isotopic enrichment for quantitative experiments.

  • Purity ≥98.0%.
  • Isotopically labeled at the bicarbonate carbon (13C).
  • Molecular formula NaH13CO3, molecular weight 85.00.
  • Available in small-scale packages including 100 mg for analytical use.
  • Recommended storage: sealed, away from moisture and light at 4°C; in solvent: -80°C up to 6 months.
  • Suitable for tracer studies, metabolic labeling, and analytical standards.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.