Learn More
Bioworld Bicarbonate Buffer, 0.05M, pH 8.0, 1 L
Catalog No. 50255427
Encompass_Preferred
Product DescriptionBicarbonate buffer has a concentration of 0.05M and a pH at 8.0. It has a boiling point of 851°C, a melting point of 300°C, and a density of 2.173 g/cm3. Bicarbonate buffering system is important in different cellular processes such as its use to maintain the pH level of mammalian blood, the formation of acid in the lumen of the stomach, and to neutralize the pH of the chyme. It has an acidity of 10.3 and is of ultrapure grade.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-255-427 Supplier Bioworld Supplier No. 402200042
Product DescriptionBicarbonate buffer has a concentration of 0.05M and a pH at 8.0. It has a boiling point of 851°C, a melting point of 300°C, and a density of 2.173 g/cm3. Bicarbonate buffering system is important in different cellular processes such as its use to maintain the pH level of mammalian blood, the formation of acid in the lumen of the stomach, and to neutralize the pH of the chyme. It has an acidity of 10.3 and is of ultrapure grade.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.