Learn More
Bioworld Bicarbonate Buffer, 0.05M, pH 9.0, 500 mL
Catalog No. 50255438
Encompass_Preferred
Bicarbonate Buffer Common buffer for immunology Technical Information:Ultrapure GradeSynonym:Sodium Hydrogen CarbonateAcidity: 10.3Appearance: LiquidBoiling Point : 851°CDensity : 2.173 g/cm3HS Code : 28363000IUPAC_Name: Sodium Hydrogen CarbonateMelting Point: >300 °C(lit.)PH Value: 9.0RTECS Number: VZ0950000
Catalog No. 50-255-438 Supplier Bioworld Supplier No. 402200081
Bicarbonate Buffer Common buffer for immunology Technical Information:Ultrapure GradeSynonym:Sodium Hydrogen CarbonateAcidity: 10.3Appearance: LiquidBoiling Point : 851°CDensity : 2.173 g/cm3HS Code : 28363000IUPAC_Name: Sodium Hydrogen CarbonateMelting Point: >300 °C(lit.)PH Value: 9.0RTECS Number: VZ0950000