Learn More
DIASYS DIAGNOSTIC SYSTEMS USA LLC DiaSys TruLab Bicarbonate is a liquid-stable and assayed quality control material for monitoring assay performance of quantitative in vitro determination of Bicarbonate (3x3mL)
Catalog No. NC2874328
Encompass_Preferred
DiaSys TruLab Bicarbonate is a set of three liquid stable controls with one level. The control is based on an aqueous solution. For veterinary or research use only. DiaSys TruLab Bicarbonate, both opened and unopened, must be stored refrigerated. Avoid contamination and protect from light. Once opened, DiaSys TruLab Bicarbonate is stable for 3 months if kept refrigerated and recapped immediatley after use.
This item is not returnable. View return policy
Catalog No. NC2874328 Supplier DIASYS DIAGNOSTIC SYSTEMS USA LLC Supplier No. 597009911065
DiaSys TruLab Bicarbonate is a set of three liquid stable controls with one level. The control is based on an aqueous solution. For veterinary or research use only. DiaSys TruLab Bicarbonate, both opened and unopened, must be stored refrigerated. Avoid contamination and protect from light. Once opened, DiaSys TruLab Bicarbonate is stable for 3 months if kept refrigerated and recapped immediatley after use.