Learn More
ALADDIN SCIENTIFIC CORPORATION Bicinchoninic acid disodium sa, 25g
Catalog No. 5000082500
Encompass_Preferred
For the determination of protein concentration Bicinchoninic acid disodium salt hydrate has been used to monitor the levels of reducing sugars thereby helping in the detection of chitinases It has also been used to determine the amount of cellulose reducing ends (micromoles/g cellulose) It has been used for determining protein concentration
Catalog No. 50-000-82500 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. B10765825G
For the determination of protein concentration Bicinchoninic acid disodium salt hydrate has been used to monitor the levels of reducing sugars thereby helping in the detection of chitinases It has also been used to determine the amount of cellulose reducing ends (micromoles/g cellulose) It has been used for determining protein concentration