Learn More
Medchemexpress LLC Bicine | 150-25-4 | 98.7% | 25 G
SDP
Catalog No. 5000414093
Encompass_Preferred
Bicine is a buffer utilized in physiological research. It is a derivative of the amino acid glycine that enhances the sensitivity of guanosine enzyme histochemical detection.
  • Buffer utilized in physiological research
  • Derivative of the amino acid glycine
  • Enhances the sensitivity of guanosine enzyme histochemical detection
Catalog No. 50-004-14093 Supplier Medchemexpress LLC Supplier No. 25GHYD0868
Bicine is a buffer utilized in physiological research. It is a derivative of the amino acid glycine that enhances the sensitivity of guanosine enzyme histochemical detection.
  • Buffer utilized in physiological research
  • Derivative of the amino acid glycine
  • Enhances the sensitivity of guanosine enzyme histochemical detection