Learn More
Bioworld Bicine, 500 g
Catalog No. 50255490
Encompass_Preferred
A zwitterionic amino acid buffer agent used for low temperature work Technical Information:Synonym: N,N-Bis(2-hydroxyethyl)glycine Titration: > 98.0% (0.1N NaOH)Water content: < 2.0%Usage: bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.Appearance: PowderBoiling Point : 388.55 °C at 760 mmHgDensity : 1.05 g/cm3 at 20 °C (lit.)HS Code : 29225000Melting Point: 190 °C (dec.)(lit.)PH Value: 4.3-5.3 (1%)RTECS Number: MB9700000
Catalog No. 50-255-490 Supplier Bioworld Supplier No. 402200962
A zwitterionic amino acid buffer agent used for low temperature work Technical Information:Synonym: N,N-Bis(2-hydroxyethyl)glycine Titration: > 98.0% (0.1N NaOH)Water content: < 2.0%Usage: bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.Appearance: PowderBoiling Point : 388.55 °C at 760 mmHgDensity : 1.05 g/cm3 at 20 °C (lit.)HS Code : 29225000Melting Point: 190 °C (dec.)(lit.)PH Value: 4.3-5.3 (1%)RTECS Number: MB9700000