Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicine, 99.38%, MP Biomedicals™

Catalog No. ICN10100580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10100580 100 g
ICN10100583 250 g
ICN10100525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10100580 Supplier MP Biomedicals, Inc Supplier No. 0210100580
Add to Cart
Add to Cart

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N, N-bis [2-hydroxyethyl]glycine
PubChem CID 8761
ChEBI CHEBI:40957
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 0.9938
IUPAC Name sodium 2-[bis(2-hydroxyethyl)amino]acetate
Formula Weight 163.2
Chemical Name or Material Bicine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.