Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals BICINE, 99%

Catalog No. AAA149570B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA149570B 1000 g
AAA1495718 50 g
AAA1495730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA149570B Supplier Thermo Scientific Chemicals Supplier No. A149570B
  • $635.50
    $517.65 / Each of 1

    Save $117.85

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol

Bicine is widely utilized as a buffering agent for low temperature work. It is used in kinetic study of a quaternary transition-state analogue complex of creatine kinase reaction. It is employed in the thin layer ion exchange chromatography method for protein resolution. It is employed in serum guanase determination by the preparation of stable substrate solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicine is widely utilized as a buffering agent for low temperature work. It is used in kinetic study of a quaternary transition-state analogue complex of creatine kinase reaction. It is employed in the thin layer ion exchange chromatography method for protein resolution. It is employed in serum guanase determination by the preparation of stable substrate solution.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Melting Point ∽191°C (decomposition)
Quantity 1000 g
Assay Percent Range 99%
Beilstein 1769362
Merck Index 14,1201
Solubility Information Soluble in water.
Formula Weight 163.17
Percent Purity 99%
Chemical Name or Material BICINE
EINECSNumber 205-755-1
RTECSNumber MB9700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.