Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Bicine, 99+%, for biochemistry

Catalog No. AC327711000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327711000 100 g Plastic bottle
AC327710250 25 g Plastic bottle
AC327715000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC327711000 Supplier Thermo Scientific Chemicals Supplier No. 327711000
  • $132.90
    $112.65 / Each of 1

    Save $20.25

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N, N-Bis(2-hydroxyethyl)glycine, Diethylolglycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Absorbance 0.05 max. (0.5M, water) at 260nm, 0.05 max. (0.5M, water) at 280nm
Melting Point 190.0°C to 192.0°C
CAS Min % 99.0
Color White
CAS Max % 100.0
Physical Form Powder
Quantity 100 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula (HOCH2CH2)2NCH2CO2H
Packaging Plastic bottle
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in ethanol, ether, chloroform and, benzene
Formula Weight 163.17
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material Bicine, For biochemistry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.