Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicine, MP Biomedicals

Catalog No. MP210100591 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210100591 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210100591 Supplier MP Biomedicals, Inc Supplier No. 0210100591
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biological buffer suitable for protein and peptide crystallization and SDS-PAGE applications.

  • Bicine is used in a thin layer ion exchange chromatography method for protein resolution
  • It is used in peptide and protein crystallization
  • Bicine is used in a kinetic study of a quaternary transition-state analogue complex of creatine kinase and in multiphasic buffer system for SDS-PAGE of proteins

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym bicine, n,n-bis 2-hydroxyethyl glycine, diethylolglycine, dihydroxyethylglycine, diethanol glycine, n,n-di 2-hydroxyethyl glycine, bicene, 2-bis 2-hydroxyethyl amino acetic acid, n,n-dihydroxyethyl glycine, n,n-dihydroxyethylglycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Melting Point 190°C (literature)
Color White
For Use With (Application) Protein Electrophoresis
Physical Form Powder
Quantity 1 kg
Solution Type Buffer
Linear Formula C6H13NO4
Solubility Information Soluble in water (10mg/mL-clear, colorless solution; a saturated solution in water is 1.1 M )
Formula Weight 163.2
Grade Research
Chemical Name or Material BICINE
Show More Show Less
Recommended Storage Store at Room Temperature (15-30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.