Learn More
Research Products International Corp Bicine Ultra-Pure, 500 Grams
SDP
Catalog No. 50488556
Encompass_Preferred
Zwitterionic buffer used in a variety of molecular biology applications such as protein resolution, protein crystallization and SDS-PAGE.
pKa @ 25°C: 8.3
pH Range: 7.6-9.0
Melting Range: 190-192°C
Catalog No. 50-488-556 Supplier Research Products International Corp Supplier No. B74060500.0
Zwitterionic buffer used in a variety of molecular biology applications such as protein resolution, protein crystallization and SDS-PAGE.
pKa @ 25°C: 8.3
pH Range: 7.6-9.0
Melting Range: 190-192°C