Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Bicine, ULTROL™ Grade, Calbiochem™,

Catalog No. 3913361KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-133-61KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-133-61KG Supplier MilliporeSigma™ Supplier No. 3913361KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A zwitterionic amino acid buffer active in the pH 7.6 to 9.0 range

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N,N-bis-(2-Hydroxyethyl)glycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name sodium 2-[bis(2-hydroxyethyl)amino]acetate
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Absorbance 5%, H2O, 250nm
pH 3.5 to 5.0
Physical Form Solid
Quantity 1 kg
Solution Type Buffer
Assay Percent Range ≥99.0%
Solubility Information Easily soluble in cold water
Formula Weight 163.2
Percent Purity >99% (titration)
Grade by titration
Chemical Name or Material zwitterionic amino acid buffer
Show More Show Less
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.