Learn More
Medchemexpress LLC HY-17605 1mg Medchemexpress, Bictegravir CAS:1611493-60-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY176051MG
Encompass_Preferred
Medchemexpress, HY-17605 1mg Bictegravir CAS:1611493-60-7 Bictegravir is a novel, potent inhibitor of HIV-1 integrase with an IC50 of 7.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9248