Learn More
TARGETMOL CHEMICALS INC BICTEGRAVIR SODIUM 5MG
SDP
Catalog No. 502580222
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Bictegravir Sodium (GS-9883 Sodium) is a potent inhibitor of HIV-1 integrase with an IC50 of 7.5 nM. Bictegravir Sodium exhibits potent and selective anti-HIV activity and low cytotoxicitypurity: 98%
Catalog No. 50-258-0222 Supplier TARGETMOL CHEMICALS INC Supplier No. T353405MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Bictegravir Sodium (GS-9883 Sodium) is a potent inhibitor of HIV-1 integrase with an IC50 of 7.5 nM. Bictegravir Sodium exhibits potent and selective anti-HIV activity and low cytotoxicitypurity: 98%