Learn More
Cayman Chemical BIctegravIrsodIum salt 10mg
SDP
Catalog No. 502915638
Encompass_Preferred
An HIV-1 integrase inhibitor (IC50 = 7.5 nM for strand transfer activity); has antiviral activity against clinical isolates of HIV-1 in PBMCs (EC50s = 0.04-1.7 nM); inhibits HIV-1 viral infection in MT-2 and MT-4 cells, CD4+ T cells, and macrophages (EC50s = 1.5, 2.5, 1.5, and 6.6 nM, respectively)
Catalog No. 50-291-5638 Supplier Cayman Chemical Supplier No. 3748810 MG
An HIV-1 integrase inhibitor (IC50 = 7.5 nM for strand transfer activity); has antiviral activity against clinical isolates of HIV-1 in PBMCs (EC50s = 0.04-1.7 nM); inhibits HIV-1 viral infection in MT-2 and MT-4 cells, CD4+ T cells, and macrophages (EC50s = 1.5, 2.5, 1.5, and 6.6 nM, respectively)