Learn More
Cayman Chemical (−)-Bicuculline methochloride, Purity: ≥98%, CAS: 53552-05-9, 50 mg
SDP
Catalog No. 502940836
Encompass_Preferred
An enantiomer of (+)-bicuculline methochloride; increases the firing rate in rat cortical neurons similar to the (+) isomer, but lacks GABA antagonist activity; inhibits sodium-independent GABA receptor binding (IC50 = 500 μM), which is approximately 100-fold less potent than the (+) isomer, but there is no stereoselectivity for sodium-dependent GABA receptor binding
Catalog No. 50-294-0836 Supplier Cayman Chemical Supplier No. 2780250 MG
An enantiomer of (+)-bicuculline methochloride; increases the firing rate in rat cortical neurons similar to the (+) isomer, but lacks GABA antagonist activity; inhibits sodium-independent GABA receptor binding (IC50 = 500 μM), which is approximately 100-fold less potent than the (+) isomer, but there is no stereoselectivity for sodium-dependent GABA receptor binding