Learn More
Apexbio Technology LLC (+)-Bicuculline 485-49-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062902
Encompass_Preferred
( )-Bicuculline is a competitive antagonist of gamma-aminobutyric acid type A (GABA A) receptors frequently employed as a pharmacological tool to elucidate the functional roles of inhibitory neurotransmission mediated by GABA A By antagonizing GABA binding this compound prevents chloride ion influx leading to reductions in neuronal inhibitory postsynaptic potentials (IPSPs) Additionally ( )-Bicuculline blocks small conductance calcium-activated potassium (SK) channels affecting neuronal excitability and firing patterns It is commonly utilized in electrophysiological studies such as patch-clamp or extracellular recordings to investigate synaptic transmission neuroplasticity mechanisms and epileptic activity models ( )-Bicuculline inhibits GABA A receptors with an IC 50 typically reported in the low micromolar range (approximately 2 5 M)
Catalog No. 50-000-62902 Supplier Apexbio Technology LLC Supplier No. N15925.1
( )-Bicuculline is a competitive antagonist of gamma-aminobutyric acid type A (GABA A) receptors frequently employed as a pharmacological tool to elucidate the functional roles of inhibitory neurotransmission mediated by GABA A By antagonizing GABA binding this compound prevents chloride ion influx leading to reductions in neuronal inhibitory postsynaptic potentials (IPSPs) Additionally ( )-Bicuculline blocks small conductance calcium-activated potassium (SK) channels affecting neuronal excitability and firing patterns It is commonly utilized in electrophysiological studies such as patch-clamp or extracellular recordings to investigate synaptic transmission neuroplasticity mechanisms and epileptic activity models ( )-Bicuculline inhibits GABA A receptors with an IC 50 typically reported in the low micromolar range (approximately 2 5 M)